Learn More
Enzo Life Sciences Minoxidil-(500mg). CAS: 38304-91-5
SDP
Supplier:  Enzo Life Sciences BMLKC1250500
Encompass_Preferred
A selective ATP-sensitive K+ (KATP) channel opener. It is metabolized to the more active sulfate in vivo. A clinically useful anti-alopecia. Antihypertensive. Stimulates renin secretion. Potent vascular smooth muscle relaxant. Alternative name: U-10858. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (29mg/ml), DMSO (6.5mg/ml) or water (2.2mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.