Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Spirits, Spectrum™ Chemical
SDP

Catalog No. 18604630 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can

Catalog No. 18-604-630

Supplier: Spectrum Chemical Manufacturing Corporation M1292500MLGL

Only null left
Add to Cart
Add to Cart

M1292, 64475-85-0

Spectrum™ Chemical Mineral Spirits , also known as white spirit or mineral turpentine, is a petroleum-derived liquid used as an organic solvent in decorating and painting. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 64475-85-0

Specifications

CAS Max % 1
Boiling Point 149°C to 213°C
Quantity 500 mL
Packaging Steel Can
UN Number UN1268
Grade Reagent
Chemical Name or Material Mineral Spirits
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.