Learn More
Sarstedt Inc Microvette 200 Capillary Blood Collection Tube Round Bottom K3 EDTA Additive 10.8 X 46.6 mm 200 µL Violet Screw Cap Polypropylene Tube
Supplier: Sarstedt Inc 20.1288.100
Preassembled end-to-end capillary tube. Cap is easy to open. Special cap design minimizes aerosol effect. Optimized tube geometry ensures thorough sample mixing. Color-coded caps and print for easy identification of additives and nominal values.Shelf life and batch number on each tube. Round bottom inner tube with flat base.