Learn More
New England Biolabs, Inc. Micrococcal Nuclease – 320000 gel units
SDP
Supplier:  New England Biolabs, Inc. M0247S
Encompass
Micrococcal nuclease is derived from Staphylococcus aureus and is a relatively non-specific endo-exonuclease. It is purified from a recombinant E. coli strain that digests double-stranded, single-stranded, circular and linear nucleic acids. The enzyme is active in the pH range of 7.0 - 10.0, with optimal activity at pH 9.2 for both RNA and DNA substrates. Cleavage preferences have been observed at sites rich in adenylate, deoxyadenylate or thymidylate. Both DNA and RNA are degraded to 3 phosphomononucleotides and dinucleotides. Micrococcal Nuclease works well with the SimpleChIP Enzymatic Chromatin IP Kit (Agarose Beads) from Cell Signaling Technology.
Catalog No. 50-994-961