Learn More
                                    BYK Gardner micro-TRI-gloss
Supplier: BYK Gardner 4563
            20, 60 and 85° in one: from high to low gloss - have the specified angle on hand. For any material: paint, plastic or even mirror-like metals. All angles measure at the same location, and the results appear simultaneously. Intelligent gloss measurement with smart communication. Excellent repeatability and inter-instrument agreement guaranteed. Intelligent auto diagnosis guarantees always accurate readings. Brilliant color display easy to read - easy to use. Statistics, Pass/Fail and Memory - ideal for QC and checks in the field. Continuous mode to check uniformity over large areas. Smart-chart software: instant QC reports with trend graph and Pass/Fail limits.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.