Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metronidazole, 99%

Catalog No. AAH6025814
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAH6025814

Supplier: Thermo Scientific Chemicals H6025814

Only null left
Add to Cart

CAS: 443-48-1 | C6H9N3O3 | 171.156 g/mol

Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

Solubility
Soluble in water (10 mg/ml at 20°C), ethanol, methanol, chloroform, and dimethyl sulfoxide.

Notes
Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-48-1
Molecular Formula C6H9N3O3
Molecular Weight (g/mol) 171.156
MDL Number MFCD00009750
InChI Key VAOCPAMSLUNLGC-UHFFFAOYSA-N
Synonym metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
PubChem CID 4173
ChEBI CHEBI:6909
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
SMILES CC1=NC=C(N1CCO)[N+](=O)[O-]

Specifications

Melting Point 159°C to 161°C
Quantity 25 g
Beilstein 611683
Solubility Information Soluble in water (10mg/ml at 20°C),ethanol,methanol,chloroform,and dimethyl sulfoxide.
Formula Weight 171.16
Percent Purity 99%
Chemical Name or Material Metronidazole
Hazard Category H350
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 207-136-1
RTECSNumber NI5600000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.