Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltitanium(IV) triisopropoxide, 1M solution in THF, AcroSeal™

Catalog No. AC428371000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy

Catalog No. AC428371000

Supplier: Thermo Scientific Chemicals 428371000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 18006-13-8 | C10H24O3Ti | 240.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18006-13-8, 109-99-9
Molecular Formula C10H24O3Ti
Molecular Weight (g/mol) 240.18
MDL Number MFCD03411233
InChI Key QBLPAZNBFFURKB-UHFFFAOYSA-N
Synonym titanium, methyltripropoxy, titanium,methyltris 2-propanolato-, t-4, methyltitanium triisopropoxide, 1m in tetrahydrofuran
PubChem CID 144064
IUPAC Name carbanide;propan-2-ol;titanium
SMILES [CH3-].CC(C)O.CC(C)O.CC(C)O.[Ti]

Specifications

CAS Min % 26.84, 70.48
CAS Max % 29.52, 73.16
Density 0.8950g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass bottle
Quantity 100 mL
Specific Gravity 0.895
Formula Weight 240.18
Chemical Name or Material Methyltitanium(IV) triisopropoxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.