Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue Solution (Methanol Solution) [for Cell Staining], TCI America™
SDP

Catalog No. M2392100ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mL

Catalog No. M2392100ML

Supplier: TCI America M2392100ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61-73-4
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.851
MDL Number MFCD00012111
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym methylene blue, methylthioninium chloride, basic blue 9, swiss blue, chromosmon, methylene blue n, methylenium ceruleum, urolene blue, bleu de methylene, methylene blue chloride
PubChem CID 6099
ChEBI CHEBI:6872
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Color Black-Blue
Quantity 100 mL
UN Number 1230
Formula Weight 319.85
Physical Form Liquid
Chemical Name or Material Methylene Blue Solution (Methanol Solution) [for Cell Staining]
EINECSNumber (5)-1995
RTECSNumber SO5600000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.