Learn More
Epigentek Inc Methylamp One-Step DNA Modification Kit (40 samples)
Supplier: Epigentek Inc P10101
The Methylamp One-Step DNA Modification Kit is a complete set of essential components which enables the experimenter to perform DNA methylation analysis using Epigentek's uniquely simplified and streamlined bisulfite method. The entire procedure can be completed within a mere 1 hour and 45 minutes and produces far superior results than any competitor kits. This kit is suitable for MS- PCR, real time MS-PCR, methylation sequencing, and pyrosequencing, as well as methylation microarray.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.