Learn More
Dimethyl trisulfide, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -68.0°C |
| Color | Yellow |
| Density | 1.1900g/mL |
| Boiling Point | 165.0°C to 175.0°C |
| Flash Point | 52°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5995 to 1.6025 |
| Linear Formula | CH3SSSCH3 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Harmful if swallowed. Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Call a POISON CENTER or doctor/physician if you feel unwell. If eye irritation persists: Get medical advice/attention. Wear protective gloves/prot |
| EINECSNumber | 222-910-9 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.