Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
| Assay Percent Range | 95% |
| Quantity | 1 g |
| IUPAC Name | methyl 4-amino-1-methylpyrrole-2-carboxylate;hydrochloride |
| Formula Weight | 190.63 |
| Percent Purity | 95% |
| Chemical Name or Material | Methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction