Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Premier™ Coccidioides EIA

Catalog No. 14910161 Shop All Meridian Bioscience Products
Click to view available options
Detectable Analytes:
C. immitis IgG, IgM
This item is not returnable. View return policy

Catalog No. 14-910-161

Supplier: Meridian Bioscience™ 603096

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Kit contains two separate conjugates to individually detect IgM and IgG antibodies if present in serum or CSF and enough breakaway microwells and prediluted reagents for 96 tests.

More accurate than traditional immunodiffusion and latex testing

  • CLIA Complexity: High
  • For detecting IgM and IgG antibodies to C. immitis in serum and cerebrospinal fluid
  • Test results within 65 minutes
  • Useful for monitoring therapy
  • Much faster and easier than complement fixation
  • Sensitivity: 97.4%; Specificity: 94.0%

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
CPT Code 86635
Clia Complexity High
Detectable Analytes C. immitis IgG, IgM
Sample Type CSF, Serum
Sensitivity 97.4%
Type Coccidioides EIA
For Use With (Application) More accurate than traditional immunodiffusion and Latex testing
Includes Two separate conjugates to individually detect IgM and IgG Antibodies if present in Serum or CSF and enough breakaway microwells and prediluted reagents for 96 Tests
Specificity 0.94
Test Time 65 min.
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.