Learn More
Med Vet International Merck Optimmune Opthalmic Ointment
Supplier: Med Vet International RXOPTI1/8OZ
Ophthalmic ointment for use in dogs only. Each gram contains 2 mg of cyclosporine, USP; petrolatum, USP; corn oil, NF; and Amerchol CAB base. Cyclosporine (cyclosporin A) is a cyclic undecapeptide metabolite of the fungus Tolypocladium inflatum gams. Sterile.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.