Learn More
Hach Company Membrane, Venting PTFE PK/18
Supplier:  Hach Company 2683318
Encompass
Replacement venting membrane for use with the Cold Vapor Mercury Test. PTFE 0.45µm pore size. 50mm Diameter hose barb fittings. Autoclavable. 18/pk
Catalog No. 50-199-6537
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.