Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melamine, 99%

Catalog No. AC125350050
Click to view available options
Quantity:
5 g
100 g
1 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum

Catalog No. AC125350050

Supplier: Thermo Scientific Chemicals 125350050

Only null left
Add to Cart

CAS: 108-78-1 | C3H6N6 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-78-1
Molecular Formula C3H6N6
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006055
InChI Key JDSHMPZPIAZGSV-UHFFFAOYSA-N
Synonym melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr
PubChem CID 7955
ChEBI CHEBI:27915
IUPAC Name 1,3,5-triazine-2,4,6-triamine
SMILES NC1=NC(N)=NC(N)=N1

Specifications

Melting Point 354°C
Color White
Density 1.5730g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 26, 245
Specific Gravity 1.573
Merck Index 15, 5880
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: 0.6g/L in ethanol (20°C),sparingly soluble in glycol,glycerol,pyridine,insoluble in ether,benzene,carbon tetrachloride
Formula Weight 126.12
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Melamine
Show More Show Less
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 203-615-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.