Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAP4K3 Monoclonal Antibody (219CT8.3.1)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537511
Description
MAP4K3 Monoclonal Antibody for Western Blot, IHC (P)
MAP4K3 (also known as GLK) is a member of the STE20 family of serine/threonine protein kinases and contains N-terminal catalytic domain and C-terminal regulatory domain. MAP4K3 is upstream of the MEKK1 target and is stimulated by UV radiation and TNF-alpha and can lead to the activation of the JNK signaling pathway. MAP4K3 can directly activate PKC-T, during TCR signaling and people with systemic lupus erythematosus show considerable enhanced MAP4K3 expression and activation of PKC-T, and the kinase IKK in T cells. MAP4K3 can act as a pro-apoptotic kinase which orchestrates activation of BAX via the concerted posttranscriptional modulation of PUMA, BAD, and BIM. The kinase activity of the encoded protein can be stimulated by UV radiation and tumor necrosis factor-alpha. The protein specifically activates the c-Jun N-terminal kinase (JNK) signaling pathway. Evidence suggests that it functions upstream of mitogen-activated protein kinase kinase 1 (MEKK1). MAP4K3 previously was referred to as RAB8-interacting protein-like 1 (RAB8IPL1), but it has been renamed mitogen-activated protein kinase kinase kinase 3 (MAP4K3).
Specifications
| MAP4K3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8IVH8 | |
| MAP4K3 | |
| MAP4K3 recombinant protein. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| 219CT8.3.1 | |
| Unconjugated | |
| MAP4K3 | |
| 4833416M01Rik; 4833416M07Rik; 9530052P13Rik; germinal center kinase-like kinase; germinal center kinase-related protein kinase; GLK; MAP4K3; MAPK/ERK kinase kinase kinase 3; MAPKKKK3; MEK kinase kinase 3; MEKKK 3; MEKKK3; mitogen activated protein kinase kinase kinase kinase 3; mitogen-activated protein kinase kinase kinase kinase 3; RAB8IPL1 | |
| Mouse | |
| Euglobulin Precipitation | |
| RUO | |
| 8491 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction