Learn More
Research Products International Corp Malt Extract Agar, 500 Grams

Supplier: Research Products International Corp M21500500.0
Malt Extract Agar is used for the isolation, cultivation and maintenance of yeasts and molds. Formulation is similar to Malt Extract Broth with the addition of agar and glycerol. This broth medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.
Dissolve 33.6 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.
Ingredients:
Dextrin: 2.75 g/L
Peptone: 0.78 g/L
Final pH (25°C): 4.6 ± 0.2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.