Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 649
Click to view available options
Quantity:
1 mg
Description
- Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 649, is a bright deep-red/far-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research.
- LEL-DyLight 649 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.
- Very bright DyLight 649 conjugate detected with traditional Cy5/deep-red filter set, Ex/Em: 655/670 nm
- Applications: immunofluorescence, glycobiology
- Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
- Inhibiting/eluting sugar: chitin hydrolysate
- Blood group specificity: non-specific
- Unconjugated lectin molecular weight: 71 KDa.
- Immunofluorescence working concentration: 5–20 μg/mL
- LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2. If a precipitate forms upon long-term storage, warm to 37°C.
Specifications
Specifications
| Quantity | 1 mg |
| Source | Tomato |
| Conjugate | DyLight 649 |
| Content And Storage | When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept. |
| Label Type | DyLight Fluors |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Form | Solution |
| Protein Subtype | Lectins |
| Product Line | DyLight |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction