Learn More
Selleck Chemical LLC LY2109761 5mg 700874-71-1
Supplier: Selleck Chemical LLC S27045MG
LY2109761 is a novel selective TGF-? receptor type I/II (T?RI/II) dual inhibitor with Ki of 38 nM and 300 nM in a cell-free assay, respectively; shown to negatively affect the phosphorylation of Smad2. LY2109761 blocks autophagy and induces apoptosis. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.