Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Process Chlorine Analyzer Reagents: Free Chlorine

Catalog No. 15399621 Shop All Tintometer Group Products
Click to view available options
Quantity:
Each

Catalog No. 15-399-621

Supplier: Lovibond™ 530210

Only null left
Add to Cart
Edge
Add to Cart

Includes

Free Chlorine Indicator Solution (473mL), Free Chlorine Buffer Solution (473mL), DPD Indicator Powder (24g)

Formulated to work with Hach™ CL17™ and other manufacturer's online chlorine analyzers

  • Used for 30 day continuous supply

Specifications

For Use With (Equipment) Hach™ CL17™ process chlorine analyzer
Includes 473mL free chlorine indicator solution, 473mL free chlorine buffer solution, 24g DPD indicator powder
Quantity Each
Type Total Chlorine Reagent Set
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.