Learn More
Medchemexpress LLC HY-121885 5mg Medchemexpress, LMD-009 CAS:950195-51-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1218855MG
Encompass_Preferred
Medchemexpress, HY-121885 5mg LMD-009 CAS:950195-51-4 LMD-009 is a selective CCR8 nonpeptide agonist. LMD-009 mediates chemotaxis, inositol phosphate accumulation, and calcium release in high potencies with EC50s from 11 to 87 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0977