Learn More
Cole-Parmer Lightnin EVIP25MIC48 High-Torque Direct-Drive Mixer, variable-speed, offset clamp, 115 VAC
Supplier:  Cole-Parmer 5032320
Encompass_Preferred
Use these batch mixers in a wide range of applications. Choose a direct-drive model for small batch mixing or high-shear applications. Mixing shafts may be cut to suit tank depth. Use the sizing guide (available in the Technical Library) to select the appropriate mixer for your application.
Catalog No. NC1134235
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.