Learn More
Cole-Parmer Extech LT300 Precision Digital Light Meter
Supplier:  Cole-Parmer 5051093
Encompass
This compact, rugged light meter features Peak mode to capture your highest readings and relative mode indicating change in light levels for added flexibility. The min/max feature stores maximum and minimum readings on the large LCD with analog bargraph. Unit measures in lux or footcandles, and provides cosine- and color-corrected measurements.
Catalog No. 50-993-067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.