Learn More
MSC Lincoln 1340 4.5 Strokes per Gal, 3/4" Outlet, 2 GPM, Brass Hand Operated Barrel Lift Pump 14 oz per Stroke, 17" Overall Length, For 15 to 55 Gallon Drums, For Paint Thinners & Petrolem Based Media
Supplier: MSC 55374177
Lincoln 1340 4.5 Strokes per Gal, 3/4" Outlet, 2 GPM, Brass Hand Operated Barrel Lift Pump 14 oz per Stroke, 17" Overall Length, For 15 to 55 Gallon Drums, For Paint Thinners & Petrolem Based Media