Learn More
Leneta Company Inc Form 5C Opacity Chart, 250/pk, 4pk/cs
Supplier: Leneta Company Inc 5C
Form 5C is a black and white sealed opacity chart. The top half is white and the bottom half is black. Overall dimensions are 7-5/8 x 10-1/4 in (194 x 260 mm). The header includes a writable area for recording: Date, Time, Formula, Batch Number, Notes. A 1/4" diameter hole is centered just under the top of the chart.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.