Learn More
Selleck Chemical LLC LDN-193189 2HCl 5mg 1435934-00-1
SDP
Supplier:  Selleck Chemical LLC S75075MG
Encompass_Preferred
LDN193189 HCl is the hydrochloride salt of LDN193189, which is a selective BMP signaling inhibitor, and inhibits the transcriptional activity of the BMP type I receptors ALK2 and ALK3 with IC50 of 5 nM and 30 nM in C2C12 cell lines, respectively, 200-fold selectivity for BMP versus TGF-?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4871
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.