Learn More
Chemie Tek LCL-161, 10MG, 1005342-46-0
Supplier: Chemie Tek CTLCL16110MG
LCL-161 is an orally bioavailable and cell permeable second mitochondrial-derived activator of caspases (SMAC) mimetic and inhibitor of IAP (Inhibitor of Apoptosis Protein) family of proteins.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.