Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LB Broth (Lennox)

Catalog No. AAH2676022
Click to view available options
Quantity:
100 g
500 g

Catalog No. AAH2676022

Supplier: Thermo Scientific Chemicals H2676022

Only null left
Add to Cart

LB Broth (Lennox), also designed as Luria-Bertani broth (Lennox) or lysogeny broth, is a nutritionally rich medium developed by Lennox for the growth and maintenance of a pure culture of recombinant strains of Escherichia coli.

LB Broth (Lennox) is nutritionally rich medium developed by Lennox for the growth and maintenance of pure culture of recombinant strains of E. coli. It is a rich growth medium that contains all the nutritional requirements for E.Coli stains. In the formula, Tryptone provides nitrogen, vitamins, minerals and amino acids essential for growths. Yeast extract is source of vitamins, particularly the B-group. Sodium chloride supplies essential electrolytes for transport and osmotic balance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• LB Broth is composed of tryptone (pancreatic digest of casein), sodium chloride, and yeast extract. The tryptone provides nitrogen, vitamins, minerals, and amino acids essential for the growth of these strains. Yeast extract is a source of vitamins, particularly the B-group. The sodium chloride supplies essential electrolytes for transport and osmotic balance

Application

• LB Broth (Lennox) is a rich growth medium that contains all the nutritional requirements for the growth and maintenance of a pure culture of recombinant Escherichia coli strains to allow for plasmid purification and/or protein expression
• LB Broth (Lennox) is used for resuspension of T. pyriformis cell fractions in a study to perform the fractionation of cellular extracts

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Packaging Type Glass Bottle
Quantity 100 g
Sensitivity Hygroscopic; Moisture sensitive

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.