Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halyard Health Lavender Nitrile™ Gloves

Catalog No. 23500122
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Catalog No. 23-500-122

Supplier: Halyard Health 52816H

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Thinner, lighter, and more economical than standard nitrile gloves, retaining protective properties

  • Powder-free
  • Ambidextrous
  • Beaded cuffs
  • Non-sterile
  • Textured fingertips
  • Higher count per box saving space and waste
  • Palm Gauge thickness 2.8mil

Certifications

510K approved for medical use

Specifications

Color Lavender
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Sterility Non-sterile
Finger Style Textured
Glove Cuff Style Beaded/Rolled
Model 52816H
Tensile Strength 24
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.