Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lasalocid A Sodium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. p-7214109
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Lasalocid is an antibacterial agent, belonging to the carboxylic polyether group of antibiotics, produced by the fermentation of Streptomyces lasaliensis. It is commercially available as a sodium salt and is reported to be used as a coccidiostat in poultry.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25999-20-6
Melting Point 175°C to 185°C; 180°C (decomposition) (literature)
Flash Point Not applicable
Molecular Formula C34H53NaO8
Linear Formula C34H53NaO8
MDL Number MFCD00078065
Quantity 1 mg
Synonym Lasalocid Sodium Salt
Optical Rotation [α]/D -28°C ± 3°C = 1 in. methanol
Formula Weight 612.77
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.