Learn More
IBI SCIENTIFIC 3P Large Mouse Restraint, 1.75" x 1.75"
Supplier: IBI SCIENTIFIC 3P MH125
Large Mouse Holder: 1.25-inch ID. IBI Scientific. Constructed of 1/4-inch clear cast acrylic, complete with adjustable nose restraint and tightening knob. Designed for mouse tail warming, injections, bleeds, and other injections.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.