Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Lab Pack Open Head Poly Drum, 30 gal.
GREENER_CHOICE

Catalog No. 19338445 Shop All Eagle Manufacturing Products
Click to view available options
Color:
Black
Blue
Red
Yellow
Closure Type:
Metal Lever-lock
Plastic Lever-lock

Catalog No. 19-338-445

Supplier: Eagle™ 1601RED

Only null left
Add to Cart
Add to Cart

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Plastic Lever-lock
Capacity (English) 30 gal.
Capacity (Metric) 113.7 L
Color Red
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 26.38 in.
Height (Metric) 67 cm
Weight (English) 18 lb.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.