Learn More
Medchemexpress LLC HY-18237 5mg Medchemexpress, KX1-004 CAS:518058-84-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY182375MG
Encompass_Preferred
Medchemexpress, HY-18237 5mg KX1-004 CAS:518058-84-9 KX1-004 is a potent and non-ATP competitive Src-PTK inhibitor with an IC50 of 40 μM. KX1-004 protects the cochlea from hazardous noise and prevents noise-induced hearing loss (NIHL). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2027