Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMT3B/NSD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP255512
Description
KMT3B/NSD1 Polyclonal specifically detects KMT3B/NSD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| KMT3B/NSD1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| Androgen receptor coactivator 267 kDa protein, Androgen receptor-associated protein of 267 kDa, ARA267STO, DKFZp666C163, EC 2.1.1.43, FLJ10684, FLJ22263, FLJ44628, H3 lysine-36 and H4 lysine-20 specific, KMT3BSotos syndrome, Lysine N-methyltransferase 3B, NR-binding SET domain-containing protein, nuclear receptor binding SET domain protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| NSD1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:INEECSLKCCSSDTKGSPLASISKSGKVDGLKLLNNMHEKTRDSSDIETAVVKHVLSELKELSYRSLGEDVSDSGTSKPSKPLLFSSASSQNHIPIEPDYKFSTLLMMLKDMHDSKTKEQRLMTAQNLVSY | |
| 100 μL | |
| Apoptosis, Breast Cancer, Cancer, Cellular Markers, Tumor Suppressors | |
| 64324 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction