Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kim-Kap™ Closures
GSA_VASDP

Catalog No. p-74480 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
For Use With (Equipment):
13 mm Plain-end Culture Tubes
16 mm Plain-end Culture Tubes
18 mm Plain-end Culture Tubes
20 mm Baffled Culture Flasks
20 mm Plain-end Culture Tubes
25 mm Plain-end Culture Tubes
38 mm Plain-end Culture Tubes
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
14-957-91C Natural 20 mm Plain-end Culture Tubes
14-957-91B Natural 18 mm Plain-end Culture Tubes
14-957-91 Natural 13 mm Plain-end Culture Tubes
14-957-91A Natural 16 mm Plain-end Culture Tubes
14-957-91D Natural 25 mm Plain-end Culture Tubes
14-957-91E Natural 38 mm Plain-end Culture Tubes
14-957-92 Red 13 mm Plain-end Culture Tubes
14-957-92A Green 13 mm Plain-end Culture Tubes
14-957-92B Yellow 13 mm Plain-end Culture Tubes
14-957-92C Blue 13 mm Plain-end Culture Tubes
14-957-92D Red 16 mm Plain-end Culture Tubes
14-957-92F Yellow 16 mm Plain-end Culture Tubes
14-957-92E Green 16 mm Plain-end Culture Tubes
14-957-92J Green 18 mm Plain-end Culture Tubes
14-957-92G Blue 16 mm Plain-end Culture Tubes
14-957-92H Red 18 mm Plain-end Culture Tubes
14-957-92L Blue 18 mm Plain-end Culture Tubes
14-957-92K Yellow 18 mm Plain-end Culture Tubes
14-957-92M Red 20 mm Plain-end Culture Tubes
14-957-92Q Blue 20 mm Plain-end Culture Tubes
14-957-92R Red 25 mm Plain-end Culture Tubes
14-957-92P Yellow 20 mm Baffled Culture Flasks
14-957-92S Green 25 mm Plain-end Culture Tubes
14-957-92T Yellow 25 mm Plain-end Culture Tubes
14-957-92N Green 20 mm Plain-end Culture Tubes
14-957-92V Blue 25 mm Plain-end Culture Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. 14-957-91C Supplier DWK Life Sciences Supplier No. 7366020
Only null left
Add to Cart
Add to Cart

Available in natural (clear) or choice of four colors

  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Push Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Sterility Non-sterile
For Use With (Equipment) 20 mm Plain-end Culture Tubes
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.