Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kemtec™ Shampoo Evaluation Kit
Shop All AquaPhoenix Scientific Products

Click to view available options
Grade:
8 to 12
Description
Includes
Evaporating dishes, pH strips, blank labels, shampoo label, jar with lid, copper shot, ink, instructor manual, student data sheets, SDS
Requires
Heat lamp or hot plate, stopwatches, lab glassware and six brands of shampoo
- Comparisons in this activity include pH, viscosity, percent solids, foam generation, foam retention and ink dispersion
- Detergent micelles and the hydrophobic and hydrophilic properties of cationic and anionic detergents are investigated
- Students perform cost analysis on the shampoos
- Grades 8-12
- Class size: 24 students
Specifications
Specifications
| Grade | 8 to 12 |
| For Use With (Application) | Make and test shampoos in the classroom |
| Includes | Evaporating dishes, ph paper, blank labels for bottles, shampoo label, plastic jars with lid, copper shot, india ink, instructor manual, student data sheets, methods of assessment manual, special needs materials, msds |
| Product Type | Shampoo Evaluation Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction