Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™: Art Forgery
SCI_ED

Catalog No. S06738 Shop All AquaPhoenix Scientific Products
Click to view available options
Grade:
6
Contains lead.

Catalog No. S06738

Supplier: Aqua Phoenix Scientific 10524

Only null left
Add to Cart
Add to Cart
Contains lead.

Includes

  • Single edge razor blades, magnifying lenses, paraffin cubes, pipets, six pairs of paintings, plastic bottles with dropper caps, lead nitrate solution, potassium chromate solution, instructor manual, student data sheets, methods of assessment manual, MSDS
  • Refill includes: paraffin cubes, six pairs of paintings, lead nitrate solution, potassium chromate solution

Requires

UV light

Introduce students to practical applications of scientific observation and reasoning.

  • Add excitement to an art, chemistry or general science class
  • Real and forged painting in the styles of Claude Monet (impressionist), Vincent Van Gogh (post-impressionist) and Jackson Pollack (abstractionist) are presented to students
  • Students then determine which painting is original and which is forgery by using physical, chemical and microscopic methods of analysis
  • Methods of analysis include testing for lead, testing for fluorescence and paint chip analysis
  • Grades 6-12
  • Class size: 24 students (six groups of four)

Specifications

Certifications/Compliance National standards
Grade 6
For Use With (Application) Used to distinguish original paintings from forgeries
Includes Instructor manual, student data sheets, msds, single edge razor blades, magnifying lenses, paraffin cubes, pipets, 6 pairs of paintings, empty dropper bottles with caps, lead nitrate solution, potassium chromate solution
Requires UV lamp, 30 small test tubes with stoppers, distilled water, spatulas and match books
Class Size 24 Students
Product Type Art Forgery Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.