Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ Piggyback Flammable Safety Cabinet
GREENER_CHOICE

Catalog No. p-27070 Shop All Justrite Safety Group Products
Click to view available options
Load Bearing Capacity:
12 gal.
17 gal.
Door Style:
Manual-close
Self-close

Catalog No. 19-121-850

Supplier: Justrite™ 891700

Only null left
Add to Cart
Add to Cart

Designed to mount on top of 30- and 45 gallon Sure-Grip™ EX cabinets, these Piggyback safety cabinets can be used independently or stacked as benchtop units for added storage.

Piggyback Safety Cabinet Features

  • Stack atop standard 30 gal. (114L) safety cabinets
  • Can also be wall mounted or used on countertop
  • 18-gauge steel with 1.5 in. (3.8cm) insulating air space
  • Available with manual or self-closing doors with flush-mounted Sure-Grip™ handles with exclusive gripper pad for secure hold and security

Features of Justrite Flammables Cabinets

  • Double-wall construction provides 1 1/2 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Backed by an exclusive 10-year warranty
  • Meet OSHA and NFPA requirements

Certifications

FM approved. Self-closing models also UL listed.

Compliance

Comply with Uniform Fire Code.

Specifications

Dimensions (D x W x H) Interior 14.56 x 39.5 x 19.5 in.
Product Type Piggyback Storage Cabinet
Load Bearing Capacity 17 gal.
Weight (Metric) 67 kg
Height (Metric) Interior 49.5 cm
Depth (Metric) Exterior 45.7 cm
No. of Shelves 1
Height (Metric) Exterior 60.9 cm
Depth (Metric) Interior 37 cm
Width (Metric) Exterior 109 cm
Width (Metric) Interior 100.3 cm
Height (English) Interior 19.5 in.
Includes Flush mounted Sure-Grip™ handles with cylinder lock and key set or an optional padlock, Haz-Alert™ reflective trilingual warning labels, dual vents with flame arresters, four adjustable self leveling feet, grounding con
No. of Doors 2
Weight (English) 148 lb.
For Use With (Application) Flammables
Width (English) Interior 39.5 in.
Depth (English) Interior 14.56 in.
Height (English) Exterior 24 in.
Color Yellow
Certifications/Compliance FM, NFPA, OSHA
Material Steel (CR)
Depth (English) Exterior 18 in.
Width (English) Exterior 43 in.
Door Style Manual-close
Surface Coating Epoxy, Polyester
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.