Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Chemcor™ Lined Safety Cabinets for Hazardous Materials
GREENER_CHOICE

Catalog No. 19414811 Shop All Justrite Safety Group Products
Click to view available options
Load Bearing Capacity:
113.5 L
170 L
227 L
No. of Shelves:
1
2

Catalog No. 19-414-811

Supplier: Justrite™ 8645282

Only null left
Add to Cart
Add to Cart
Request a Quote

Give your workers the edge. Justrite ChemCor™ thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals.

  • Protection against corrosion from drips and off-gassing from harsh chemicals
  • Feature unique, flame-coated thermoplastic finish on interior doors, shelves, walls, ceiling, and sump
  • Non-porous, seam-free surface eliminates need for corrosion-prone rivets or fasteners
  • Resists staining and cleans up easily
  • Includes separate polyethylene work tray that can be placed on top

Specifications

Load Bearing Capacity 170 L
Product Type Lined Safety Cabinet
Door Style Self-close
For Use With (Application) Hazardous materials
No. of Doors 2
No. of Shelves 2
Color Blue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.