Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Itaconic acid, 99+%

Catalog No. AC122812500
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122812500 250 g Plastic bottle
AC122810010 1 kg Plastic bottle
AC122810051 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC122812500 Supplier Thermo Scientific Chemicals Supplier No. 122812500
Only null left
Add to Cart

CAS: 97-65-4 | C5H6O4 | 130.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-65-4
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.1
InChI Key LVHBHZANLOWSRM-UHFFFAOYSA-N
Synonym itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid
PubChem CID 811
ChEBI CHEBI:30838
IUPAC Name 2-methylidenebutanedioic acid
SMILES C=C(CC(=O)O)C(=O)O

Specifications

Melting Point 165.0°C to 168.0°C
Color Beige to White
Density 1.5700g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula HO2CCH2C(=CH2)CO2H
Quantity 250 g
Beilstein 02,76
Merck Index 15,529
Specific Gravity 1.57
Solubility Information Solubility in water: 95g/L (25°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in most other common organic,solvents
Formula Weight 130.1
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Itaconic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 202-599-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.