Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Isotemp™ 400 Refrigerated Incubator, 381 L, Stainless Steel
Shop All Fisherbrand Products

Click to view available options
Capacity (Metric):
381 L
Description
These Isotemp units use a Peltier device which can both heat and cool precisely to the desired temperature.
Benefits of Peltier technology:
Key Features of Isotemp Peltier Low Temperature Incubators:
Benefits of Peltier technology:
- Low energy consumption, especially in temperature range 15 to 25 ºC
- No hazardous refrigerants
- No bulky compressor with random vibrations
- Excellent temperature stability and uniformity
Key Features of Isotemp Peltier Low Temperature Incubators:
- Access port with silicone plug for independent data logging or for use of electrical equipment inside the unit
- Peltier device provides precise temperature within chamber – from 10° to 60°C
- Large easy-to-view fluorescent display with intuitive microprocessor controller
- Internal glass door for undisturbed sample viewing
- Readily stackable same-size units without the need for tools or kits
- Flexible shelving system with different positions to optimize chamber space
- Lockable casters for easy relocation (large unit)
- Outstanding temperature uniformity and stability to keep samples at desired condition
- Wide temperature range: 10° to 60°C
- Intuitive and easy-to-use microprocessor controller
- Automatic over temperature alarm – no operator settings required
- Heated door prevents condensation at lower temperatures
- Timer function for pre-set power-up or shut off (up to 99 hours)
- Corrosion resistant stainless steel* interior
- Easy-to-clean rounded interior edges, designed to reduce contamination risks
- Flexible shelving with no tools required to set up, adjust or remove
- Small external dimensions per chamber size reduce required bench / floor space
- Standard RS232 interface for datalogging (no software provided)
- 2 year parts and labor warranty
- cCSAus certified
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (English) | 13.5 cu. ft. |
| Capacity (Metric) | 381 L |
| Chamber Material | Stainless Steel |
| No. of Shelves | 2 supplied/17 max. |
| Plug Type | Nema 5-15 US |
| Temperature Range (Metric) | 10°C to 60°C |
| Type | Refrigerated Incubator |
| Model | Isotemp 400 |
| Temperature Stability | ≤ ±0.1 at 20°C |
| Temperature Uniformity | ≤ ±0.4 at 20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction