Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Laurate 97.0+%, TCI America™
SDP

Catalog No. L001425ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Catalog No. L001425ML

Supplier: TCI America L001425ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6309-51-9
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00048429
InChI Key FVKRIDSRWFEQME-UHFFFAOYSA-N
Synonym isoamyl laurate, isopentyl laurate, dodecanoic acid, 3-methylbutyl ester, lauric acid, isopentyl ester, isoamyl dodecanoate, isopentyl dodecanoate, lauric acid isoamyl ester, isopentyl dodecylate, unii-m1slx00m3m, iso-amyl n-dodecanoate
PubChem CID 61386
ChEBI CHEBI:87343
IUPAC Name 3-methylbutyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCC(C)C

Specifications

Color Colorless
Boiling Point 170°C
Quantity 25 mL
Formula Weight 270.46
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Laurate
EINECSNumber (2)-0798
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.