Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) sulfate concentrate, Honeywell™

Catalog No. 6002658 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 Pc.

Catalog No. 60-026-58

Supplier: Honeywell Chemicals 380471EA

Only null left
Add to Cart
Add to Cart

for 1L standard solution, 0.1 M FeSO4

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-21-4
MDL Number MFCD00011014

Specifications

Quantity 1 Pc.
UN Number NONH for all modes of transport
Chemical Name or Material Iron(II) sulfate concentrate
Hazard Statement P305 + P351 + P338
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.