Learn More
Specifications
Specifications
| Color | Gray |
| Melting Point | 1535°C |
| CAS | 7439-89-6 |
| Boiling Point | 3000°C |
| Atomic Weight | 55.85 |
| Density | 7.86g/mL |
| Quantity | 500 g |
| Identification | Pass test |
| Mesh Size | Finer than 100 Mesh |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 4.1, : Flammable Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.