Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Irgarol™, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 mg
Description
Agriculture
Specifications
Specifications
| Product Line | Irgarol |
| CAS | 9004-57-3 |
| Physical Form | Neat |
| Linear Formula | C11H19N5 S |
| MDL Number | MFCD01863779 |
| Synonym | 2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-s-triazine |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 253.37 |
| Grade | Analytical Standard |
Irgarol is a registered trademark of Ciba Specialty Chemicals Corp.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction