Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion exchanger Dowex™ 50 WX-8, H+ form, EMD-Millipore

Catalog No. M1052210250 Shop All MilliporeSigma Products
Click to view available options
Quantity:
250 g

Catalog No. M1052210250

Supplier: MilliporeSigma 1.05221.0250

Only null left
Add to Cart
Add to Cart

Strongly acid cation exchangers

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69011-20-7
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
Synonym porapak p, lewatit c267 ion exchange resin, ion exchange resin, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, cation exchange resin, ethidium bromide adsorber, divinylbenzene, ethylvinylbenzene, vinylbenzene copolymer, ethenylbenzene, diethenylbenzene, ethylethenylbenzene polymer, ethylbenzene, ethylethenylbenzene, diethenylbenzene polymer, sulfonated, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, sulfonated
PubChem CID 172244
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *

Specifications

Density Density: 1.040g/cm3 (20°)
Quantity 250 g
Chemical Name or Material Ion exchanger Dowex™ 50 WX 8
Recommended Storage Store at 15° to 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.