Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Ethidium Bromide, 10 mg/mL

Catalog No. 15585011
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-585-011 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-585-011 Supplier Invitrogen™ Supplier No. 15585011
Only null left
Add to Cart
Edge
Add to Cart

Includes

10mL of 10mg/mL solution

Sensitive fluorescent dye detects nucleic acids in agarose gels and cesium chloride gradients

UltraPure Ethidium Bromide is available as an aqueous solution containing 10mg/mL UltraPure ethidium bromide and high-purity water. Sensitive: Detects as little as little as 1ng of nucleic acids in agarose gels.

  • Versatile: Detects DNA or RNA
  • Easy to Use: Can be used undiluted or diluted to the desired concentration
  • An aromatic compound with a phenanthridine core, ethidium bromide has excitation maxima at 300nm and 520nm and an emission maximum at 600nm

Agarose Gel Electrophoresis, DNA & RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10ml of 10mg⁄ml solution. Store at 15°C to 30°C
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Ethidium bromide
Quantity 10 mL
Shipping Condition Room Temperature
Target Molecule RNA, DNA
Product Line UltraPure
Product Type Ethidium Bromide
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.