Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ PCR Purification Kit

Catalog No. K310001
Encompass_Preferred
Click to view available options
Quantity:
250 Preps
50 Preps

Catalog No. K310001

Supplier: Invitrogen™ K310001

Only null left
Add to Cart
Add to Cart

The PureLink PCR Purification Kit provides a more convenient, rapid, and high-yielding purification process compared to other companies' kits.

The PureLink PCR Purification Kit provides a more convenient, rapid, and high-yielding purification process compared to other companies' kits. After PCR amplification, purification of the PCR amplicons is required for many common downstream applications, including enzyme digestion, ligation, sequencing, and labeling experiments, to prevent enzymes (polymerase, restriction enzyme) and reaction components (dNTPs, primers, buffers, salts) from carrying over, contaminating, and negatively influencing the downstream experiments.

Advantages of the PureLink PCR Purification Kit include:

  • 4X binding capacity compared to Company Q; 40 μg for the PureLink kit versus 10 μg from other companies' kits
  • No need for tedious sample pH adjustments necessary with other kits
  • Convenient and rapid protocol, completed in less than 10 minutes
  • Optional size-selection buffers included for selectively purifying 300–600 bp fragments

The PureLink PCR Purification Kit is based on the selective binding of dsDNA to silica-based membrane in the presence of chaotropic salts. Simply mix the PCR product with Binding Buffer and apply to the PureLink Spin Column. The dsDNA binds to the silica-based membrane in the column. Remove impurities by thorough washing with Wash Buffer. To purify the DNA, elute the dsDNA in low-salt Elution Buffer or water.The purified PCR product is suitable for automated fluorescent DNA sequencing, restriction enzyme digestion, and cloning.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Format Kit
High-throughput Compatibility High-throughput Compatible
Product Line PureLink
Product Type PCR Purification Kit
Quantity 50 Preps
Run Time 10 min.
Shipping Condition Room Temperature
Target DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application) Next-Generation Sequencing (NGS), Cloning
Isolation Technology Silica Spin Column
Sample Type PCR Products
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.