Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ HiPure Plasmid Midiprep Kit

Catalog No. p-4926487
Encompass_Preferred
Click to view available options
Quantity:
25 Preps
50 Preps

Catalog No. K210004

Supplier: Invitrogen™ K210004

Only null left
Add to Cart
Edge
Add to Cart

The PureLink HiPure Plasmid Midiprep Kit is designed to isolate transfection-grade plasmid DNA from E. coli.

The PureLink™ HiPure Plasmid Midiprep Kit is designed to isolate transfection-grade plasmid DNA from E. coli. The Midiprep Kit protocol will typically yield 100–350 μg plasmid DNA from 15–25 mL bacterial culture, at a purity that is comparable to that achieved by two passes through cesium chloride gradients.

Advantages of the PureLink™ HiPure Plasmid Midiprep Kit include:

  • Labor and time saving—no need for extra steps to remove endotoxin and other contaminants
  • High-purity, low-endotoxin product—pure enough for mammalian cell transfection
  • Versatility—purify any type and size of plasmid DNA, including BAC, bacmids, and ssM13 DNAs

Anion-exchange chromatography for plasmid purification

The PureLink™ HiPure Plasmid DNA Purification Kits use a patented anion-exchange resin to purify plasmid DNA to a level equivalent to two passes through CsCl gradients. The resin combines excellent capacity with a fast flow rate, high resolution, high yield, and efficient endotoxin removal. Plasmid preparations can typically be completed in less than 2 hours.

Contaminant-free DNA

Unlike cesium chloride gradient protocols, PureLink™ HiPure Plasmid Purification Systems do not use organic solvents, ethidium bromide, or cesium chloride, all of which are troublesome to work with and to dispose of. Typically, plasmid DNA prepared using the PureLink™ HiPure Plasmid Midiprep Kit will have an A260/A280 ratio of greater than 1.80, indicating that the DNA is reasonably free of proteins that might interfere with downstream applications. Endotoxin levels are typically within 0.1–1 EU/μg, making this plasmid DNA ideal for mammalian cell transfection.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 100 mL Resuspension Buffer (R3)
• 550 μL RNase A
• 100 mL Lysis Buffer (L7)
• 100 mL Precipitation Buffer (N3)
• 250 mL Equilibration Buffer (EQ1)
• 500 mL Wash Buffer (W8)
• 125 mL Elution Buffer (E4)
• 15 mL TE Buffer
• 25 HiPure Columns
• 5 Column Holders

Store all components at room temperature.
Format Column
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
Elution Volume 5 mL
Final Product Type BAC DNA, Plasmid DNA
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 25 Preps
Plasmid <40kb, Low Copy Plasmid, High Copy Plasmid, BAC
Prep Scale 100-200 μg (Medium-Scale) Plasmid DNA
Product Line PureLink
Product Type Plasmid MidiPrep Kit
Quantity 25 Preps
Purity Transfection Grade
Shipping Condition Room Temperature
Target BAC DNA, Plasmid DNA
System Type PureLink™
Test Time 2 hr.
Yield 100 to 350 μg
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.