Learn More
Invitrogen™ Dynabeads™ Untouched™ Mouse CD4 Cells Kit
Description
Includes
20mL Mouse Depletion Dynabeads and 2mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119. These reagents are sufficient to process approx. 1 x 109 cells
Dynabeads Untouched Mouse CD4 Cells supplies both 4.5μm superparamagnetic Dynabeads (with a polyclonal sheep anti-rat IgG antibody covalently bound to the bead surface) and a monoclonal Antibody Mix, together designed for the depletion of CD4-negative leucocytes from mouse spleen or lymph node single-cell suspensions. Following the depletion procedure, the negatively isolated, untouched target mouse CD4+ cells are pure, viable, and ready to be used in any downstream application.
- Rapid isolation of untouched mouse CD4
- T cells—no columns required
- Pure and viable mouse CD4+ T cells are ready for any downstream application
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Type | T Cells (CD4) |
| Content And Storage | Contains: 20 mL Mouse Depletion Dynabeads™ and 2 mL Antibody Mix with monoclonal antibodies towards CD8, CD11b, CD16/32, CD45R and Ter-119. These reagents are sufficient to process ∼1 x 109 cells. Store kit components at 2°C to 8°C. |
| Product Type | Cell Isolation Kit |
| Isolation Technology | Negative Isolation |
| No. of Cells | Processes ∼1 x 109 cells total |
| Output Viability | >95% |
| Product Line | DYNAL, Dynabeads |
| Purity or Quality Grade | Research Grade |
| Quantity | 2 x 10 mL |
| Reactivity | Mouse |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.